Learn More
Sino Biological Recombinant Cynomolgus IL1RAP/IL-1RAcP Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 90934C02H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus IL1RAP (NP_001028132.1) (Met1-Glu359) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-2646