Learn More
Sino Biological Cynomolgus IL1RL1 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 90915C08H1
Encompass_Preferred
A DNA sequence encoding the cynomolgus IL1RL1 (XP_005575214.1) (Met1-Cys331) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-2290