Learn More
Sino Biological Cynomolgus IL20RB Protein 50ug (Sino Biological)
Supplier:  Sino Biological 90308K08H50
Encompass_Preferred
A DNA sequence encoding the cynomolgus IL20RB (XP_005545913.1) (Asp30-Ala230) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-2897