Learn More
Sino Biological Anti-IL-21 Receptor/IL-21R Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 90315T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Cynomolgus IL-21R / Il21R (Catalog#90315-C08H; XP_005591578.1; Met1-Pro236). IL-21R / Il21R specific IgG was purified by Cynomolgus IL-21R / Il21R affinity chromatography.
Catalog No. 50-241-0990