Learn More
Sino Biological Recombinant Cynomolgus CD25/IL-2R alpha Protein (ECD, His & AVI Tag), Avi-tag Biotinylated, HPLC-verified
Catalog No. 502993051
Encompass_Preferred
A DNA sequence encoding the Cynomolgus IL2RA (H6WS54) (Met1-Arg213) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-299-3051 Supplier Sino Biological Supplier No. 90975C49HB100
A DNA sequence encoding the Cynomolgus IL2RA (H6WS54) (Met1-Arg213) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.