Learn More
Sino Biological Recombinant Cynomolgus CD30 Ligand/TNFSF8 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 90091C07H1
Encompass_Preferred
A DNA sequence encoding the cynomolgus TNFSF8 (G7P2F1) (Gln63-Asp234) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-222-2725