Learn More
Sino Biological Cynomolgus IL3Ra Protein 1MG (Sino Biological)
Supplier:  Sino Biological 90998C08H1
Encompass_Preferred
A DNA sequence encoding the Cynomolgus IL3Ra (G8F3K3) (Met1-Arg302) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-2872