Learn More
Sino Biological Recombinant Cynomolgus IL-4R Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 90897C02H50
Encompass_Preferred
A DNA sequence encoding the cynomolgus IL4R (EHH60265.1) (Met1-Arg232) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-2661