Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Cynomolgus integrin α4β7 heterodimer protein, his-tagged α4 / tag-free β7, 100 μg

Catalog No. NC3792395
Encompass_Preferred

Recombinant cynomolgus integrin α4β7 heterodimer produced by co-expression of the α4 and β7 subunits in human HEK293 cells. MALS-verified and supplied lyophilized for use in binding assays, ELISA, and SPR bioactivity studies.

  • Expression system: human 293 (HEK293) cells.
  • Tagging: his tag on α4, β7 tag free.
  • Format: lyophilized, 100 μg vial.
  • Purity: >90% by SDS-PAGE and SEC-MALS.
  • Molecular weight: heterodimer observed ~200-250 kDa by SEC-MALS.
  • Endotoxin: <1.0 EU/μg (LAL/rFC).
  • Storage: long-term lyophilized at -20°C or below; stable 12 months.

Catalog No. NC3792395 Supplier ACROBiosystems Supplier No. IT7C52W4100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant cynomolgus integrin α4β7 heterodimer produced by co-expression of the α4 and β7 subunits in human HEK293 cells. MALS-verified and supplied lyophilized for use in binding assays, ELISA, and SPR bioactivity studies.

  • Expression system: human 293 (HEK293) cells.
  • Tagging: his tag on α4, β7 tag free.
  • Format: lyophilized, 100 μg vial.
  • Purity: >90% by SDS-PAGE and SEC-MALS.
  • Molecular weight: heterodimer observed ~200-250 kDa by SEC-MALS.
  • Endotoxin: <1.0 EU/μg (LAL/rFC).
  • Storage: long-term lyophilized at -20°C or below; stable 12 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.