Learn More
ACROBiosystems Cynomolgus ITGA7X1BITGB1 Hete
Catalog No. 502534559
Encompass_Preferred
Cynomolgus ITGA7X1BITGB1 Heterodimer Protein Tag FreeHis Tag (IT1-C52W3) is expressed from human 293 cells (HEK293). It contains AA Phe 34 - Pro 1081 (ITGA7X1B) Gln 21 - Asp 728 (ITGB1) (Accession # XP_005571182.1 (ITGA7X1B) XP_015311227.1 (ITGB1)).
Catalog No. 50-253-4559 Supplier ACROBiosystems Supplier No. IT1C52W3100UG
Cynomolgus ITGA7X1BITGB1 Heterodimer Protein Tag FreeHis Tag (IT1-C52W3) is expressed from human 293 cells (HEK293). It contains AA Phe 34 - Pro 1081 (ITGA7X1B) Gln 21 - Asp 728 (ITGB1) (Accession # XP_005571182.1 (ITGA7X1B) XP_015311227.1 (ITGB1)).