Learn More
Sino Biological Recombinant Cynomolgus ITPRIPL1 Protein (ECD, His Tag), HPLC-verified
Catalog No. 502993088
Encompass_Preferred
A DNA sequence encoding the Cynomolgus ITPRIPL1 (XP_005575041.3) (His25-Gly103) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-3088 Supplier Sino Biological Supplier No. 91051C08H100
A DNA sequence encoding the Cynomolgus ITPRIPL1 (XP_005575041.3) (His25-Gly103) was expressed with a polyhistidine tag at the C-terminus.