Learn More
Sino Biological Recombinant Cynomolgus VEGFR2/KDR Protein (ECD, His & AVI Tag), Avi-tag Biotinylated, HPLC-verified
Catalog No. 502992728
Encompass_Preferred
A DNA sequence encoding the Cynomolgus KDR (EHH53800.1) (Met1-Glu764) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-299-2728 Supplier Sino Biological Supplier No. 90907C49HB20
A DNA sequence encoding the Cynomolgus KDR (EHH53800.1) (Met1-Glu764) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.