Learn More
Sino Biological Recombinant Cynomolgus VEGFR2/KDR Protein (ECD, His Tag)
Supplier:  Sino Biological 90907C08H50
Encompass_Preferred
A DNA sequence encoding the cynomolgus KDR (EHH53800.1) (Met1-Glu764) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-2657