Learn More
Sino Biological Recombinant Cynomolgus KIR3DL3 Protein (His Tag)
Catalog No. 502992819
Encompass_Preferred
A DNA sequence encoding the Cynomolgus KIR3DL3 (XP_005590394.1) (Met1-Leu339) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-2819 Supplier Sino Biological Supplier No. 90983C08H100
 Restricted Item
Add to Cart
Add to Cart
A DNA sequence encoding the Cynomolgus KIR3DL3 (XP_005590394.1) (Met1-Leu339) was expressed with a polyhistidine tag at the C-terminus.