Learn More
ACROBiosystems Cynomolgus KLRG1 100ug
Catalog No. 502534542
Encompass_Preferred
Cynomolgus KLRG1 Protein His Tag (KL1-C5243) is expressed from human 293 cells (HEK293). It contains AA Leu 60 - Pro 189 (Accession # A0A2K5WAP9).
Catalog No. 50-253-4542 Supplier ACROBiosystems Supplier No. KL1C5243100UG
Cynomolgus KLRG1 Protein His Tag (KL1-C5243) is expressed from human 293 cells (HEK293). It contains AA Leu 60 - Pro 189 (Accession # A0A2K5WAP9).