Learn More
Sino Biological Recombinant Cynomolgus, Rhesus LIF Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 91002C49HB20
Encompass_Preferred
A DNA sequence encoding the Cynomolgus / Rhesus LIF (A0A2K5UCA6)(Ser27-Phe206) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-240-8779