Learn More
ACROBiosystems ACROBiosystems Cynomolgus LILRB2 / CD85d / ILT4 Protein, His Tag
Catalog No. 502106892
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus LILRB2, His Tag (LI2-C52H3) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Leu 448 (Accession # BAE87348.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 54.7 kDa.
Catalog No. 50-210-6892 Supplier ACROBiosystems Supplier No. LI2C52H3100UG
Recombinant Protein;100UG;Cynomolgus LILRB2, His Tag (LI2-C52H3) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Leu 448 (Accession # BAE87348.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 54.7 kDa.