Learn More
ACROBiosystems ACROBiosystems Cynomolgus LILRB4 / CD85k / ILT3 Protein, Fc Tag
Catalog No. 502106587
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus LILRB4, Fc Tag (CDK-C5258) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Glu 259 (Accession # XP_015297198.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 52.7 kDa. As a result of glycosylation, the protein migrates as 60-66 kDa under reducing (R) condition, and 120-140 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-210-6587 Supplier ACROBiosystems Supplier No. CDKC5258100UG
Recombinant Protein;100UG;Cynomolgus LILRB4, Fc Tag (CDK-C5258) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Glu 259 (Accession # XP_015297198.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 52.7 kDa. As a result of glycosylation, the protein migrates as 60-66 kDa under reducing (R) condition, and 120-140 kDa under non-reducing (NR) condition (SDS-PAGE).