Learn More
ACROBiosystems ACROBiosystems Cynomolgus LIV-1 / SLC39A6 Protein, His Tag
Catalog No. 502106602
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus LIV-1, His Tag (LV1-C52H5) is expressed from human 293 cells (HEK293). It contains AA Leu 21 - Ile 309 (Accession # XP_005586923.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 34.5 kDa. The protein migrates as 50-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus LIV-1, His Tag (Cat. No. LV1-C52H5) at 1 μg/mL (100 μL/well) can bind Anti-LIV-1 Antibody, Human IgG1 with a linear range of 0.2-2 ng/mL (QC tested).
Catalog No. 50-210-6602 Supplier ACROBiosystems Supplier No. LV1C52H5100UG
Recombinant Protein;100UG;Cynomolgus LIV-1, His Tag (LV1-C52H5) is expressed from human 293 cells (HEK293). It contains AA Leu 21 - Ile 309 (Accession # XP_005586923.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 34.5 kDa. The protein migrates as 50-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus LIV-1, His Tag (Cat. No. LV1-C52H5) at 1 μg/mL (100 μL/well) can bind Anti-LIV-1 Antibody, Human IgG1 with a linear range of 0.2-2 ng/mL (QC tested).