Learn More
ACROBiosystems Cynomolgus LTBR 100ug
Catalog No. 502534500
Encompass_Preferred
Cynomolgus LTBR / TNFRSF3 Protein His Tag (LTR-C52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 28 - Thr 229 (Accession # A0A2K5VGQ6).
Catalog No. 50-253-4500 Supplier ACROBiosystems Supplier No. LTRC52H3100UG
Cynomolgus LTBR / TNFRSF3 Protein His Tag (LTR-C52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 28 - Thr 229 (Accession # A0A2K5VGQ6).