Learn More
ACROBiosystems Cynomolgus LY6G6D 100ug
Catalog No. 502534489
Encompass_Preferred
Cynomolgus LY6G6D Protein His Tag (LYD-C5248) is expressed from human 293 cells (HEK293). It contains AA Asn 20 - Ser 104 (Accession # WAK97502.1).
Catalog No. 50-253-4489 Supplier ACROBiosystems Supplier No. LYDC5248100UG
Cynomolgus LY6G6D Protein His Tag (LYD-C5248) is expressed from human 293 cells (HEK293). It contains AA Asn 20 - Ser 104 (Accession # WAK97502.1).