Learn More
ACROBiosystems Recombinant Protein;Cynomolgus M-CSF R / CSF1R / CD115 Protein;HEK293;100UG;CSR,CSF1R,C-FMS,CD115,CSFR,FIM2,FMS,M-CSFR,CSF-1R,CSF1 R
Supplier:  ACROBiosystems CSRC52E1100UG
Encompass_Preferred
Recombinant Protein;Cynomolgus M-CSF R / CSF1R / CD115 Protein;100UG;Cynomolgus M-CSF R, His Tag (CSR-C52E1) is expressed from human 293 cells (HEK293). It contains AA Ile 99 - Pro 596 (Accession # G7P8P1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 57.1 kDa. The protein migrates as 70-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-113-8329