Learn More
ACROBiosystems Cynomolgus MARCO Protein, His Tag
Supplier: ACROBiosystems MARC5248100UG
Recombinant Protein;100UG;Cynomolgus MARCO, His Tag (MAR-C5248) is expressed from human 293 cells (HEK293). It contains AA Gln 69 - Ile 520 (Accession # A0A2K5X0Z2-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 46.7 KDa.