Learn More
ACROBiosystems Cynomolgus MARCO Protein, His Tag
Catalog No. 502054853
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus MARCO, His Tag (MAR-C5248) is expressed from human 293 cells (HEK293). It contains AA Gln 69 - Ile 520 (Accession # A0A2K5X0Z2-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 46.7 KDa.
Catalog No. 50-205-4853 Supplier ACROBiosystems Supplier No. MARC52481MG
Recombinant Protein;1MG;Cynomolgus MARCO, His Tag (MAR-C5248) is expressed from human 293 cells (HEK293). It contains AA Gln 69 - Ile 520 (Accession # A0A2K5X0Z2-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 46.7 KDa.