Learn More
ACROBiosystems ACROBiosystems Cynomolgus MCAM / CD146 Protein, His Tag (MALS verified)
Catalog No. 502106492
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus MCAM, His Tag (MCM-C52H3) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Gly 559 (Accession # A0A5F8A4A1-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 61.5 kDa. The protein migrates as 80-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6492 Supplier ACROBiosystems Supplier No. MCMC52H3100UG
Recombinant Protein;100UG;Cynomolgus MCAM, His Tag (MCM-C52H3) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Gly 559 (Accession # A0A5F8A4A1-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 61.5 kDa. The protein migrates as 80-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.