Learn More
ACROBiosystems ACROBiosystems Cynomolgus MERTK / Mer Protein, Fc Tag
Catalog No. 502106404
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus MERTK, Fc Tag (MEK-C5253) is expressed from human 293 cells (HEK293). It contains AA Ala 23- Ile 507 (Accession # XP_005575320.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 79.1 Kda.
Catalog No. 50-210-6404 Supplier ACROBiosystems Supplier No. MEKC52531MG
Recombinant Protein;1MG;Cynomolgus MERTK, Fc Tag (MEK-C5253) is expressed from human 293 cells (HEK293). It contains AA Ala 23- Ile 507 (Accession # XP_005575320.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 79.1 Kda.