Learn More
ACROBiosystems Cynomolgus MICA 100ug
Catalog No. 502534448
Encompass_Preferred
Cynomolgus MICA Protein His Tag (MIA-C52H5) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Trp 304 (Accession # A0A7N9D4J5).
Catalog No. 50-253-4448 Supplier ACROBiosystems Supplier No. MIAC52H5100UG
Cynomolgus MICA Protein His Tag (MIA-C52H5) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Trp 304 (Accession # A0A7N9D4J5).