Learn More
Sino Biological Recombinant Cynomolgus MICA Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 90924C08H1
Encompass_Preferred
A DNA sequence encoding the cynomolgus MICA (AAO24115.1) (Glu1-Pro288) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6949