Learn More
Complement Technology Inc Cyno C3b, 100 ug
Supplier:  Complement Technology Inc CY114
Encompass
Cynomolgus monkey C3b (cyno C3b) is derived from native purified cynomolgus monkey C3 (cyno C3, CompTech #CY113) upon cleavage and release of C3a with the alternative pathway C3 convertase.
Catalog No. NC1845902