Learn More
Kingfisher Biotech Inc Cynomolgus Monkey CCL2 Recombinant Protein (Yeast, Endotoxin - Free) - 100 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1284Y100
Encompass
The Cynomolgus Monkey CCL2 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Cynomolgus Monkey CCL2 applications are for cell culture, ELISA standard, and Western Blot Control. Cynomolgus Monkey CCL2 yeast-derived recombinant protein can be purchased in multiple sizes. Cynomolgus Monkey CCL2 Specifications: (Molecular Weight: 8.7 kDa) (Amino Acid Sequence: QPDAINAPVT CCYNFTNRKI SVQRLASYRR ITSSKCPKEA VIFKTIVAKE ICADPKQKWV QDSMDHLDKQ IQTPKP (76)) (Gene ID: 102135739). For research use only. Made in the USA
Catalog No. 50-253-3436