Learn More
Cell Biologics Cynomolgus Monkey Primary Bronchial Epithelial Cells, Vial
SDP
Catalog No. NC1891979
Encompass_Preferred
Cynomolgus Monkey Primary Bronchial Epithelial Cells can be used for the assay of cell-cell interaction, adhesion, PCR, Western blot, immunoprecipitation, immunofluorescent flow cytometry, or generating cell derivatives for desired research applications.
Catalog No. NC1891979 Supplier Cell Biologics Supplier No. MK6033BVIAL
Cynomolgus Monkey Primary Bronchial Epithelial Cells can be used for the assay of cell-cell interaction, adhesion, PCR, Western blot, immunoprecipitation, immunofluorescent flow cytometry, or generating cell derivatives for desired research applications.