Learn More
Kingfisher Biotech Inc Cynomolgus Monkey CXCL1 Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1865Y025
Encompass
The Cynomolgus Monkey CXCL1 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Cynomolgus Monkey CXCL1 applications are for cell culture, ELISA standard, and Western Blot Control. Cynomolgus Monkey CXCL1 yeast-derived recombinant protein can be purchased in multiple sizes. Cynomolgus Monkey CXCL1 Specifications: (Molecular Weight: 7.9 kDa) (Amino Acid Sequence: ASVVTELRCQ CLQTLQGIHP KNIQSVNVKA PGPHCAETEV IATLKNGQKA CLNPASPMVQ KIIKKMLNCD KSN (73)) (Gene ID: 574222). For research use only. Made in the USA
Catalog No. 50-253-2680