Learn More
Kingfisher Biotech Inc Cynomolgus Monkey IL-10 Recombinant Protein (Yeast, Endotoxin - Free) - 5 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1359Y005
Encompass
The Cynomolgus Monkey IL-10 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Cynomolgus Monkey IL-10 applications are for cell culture, ELISA standard, and Western Blot Control. Cynomolgus Monkey IL-10 yeast-derived recombinant protein can be purchased in multiple sizes. Cynomolgus Monkey IL-10 Specifications: (Molecular Weight: 18.7 kDa) (Amino Acid Sequence: SPGQGTQSEN SCTRFPGNLP HMLRDLRDAF SRVKTFFQMK DQLDNILLKE SLLEDFKGYL GCQALSEMIQ FYLEEVMPQA ENHDPDIKEH VNSLGENLKT LRLRLRRCHR FLPCENKSKA VEQVKNAFSK LQEKGVYKAM SEFDIFINYI EAYMTMKIQN (170)) (Gene ID: 102133450). For research use only. Made in the USA
Catalog No. 50-253-3330