Learn More
Kingfisher Biotech Inc Cynomolgus Monkey IL-22 Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1285Y025
Encompass
The Cynomolgus Monkey IL-22 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Cynomolgus Monkey IL-22 applications are for cell culture, ELISA standard, and Western Blot Control. Cynomolgus Monkey IL-22 yeast-derived recombinant protein can be purchased in multiple sizes. Cynomolgus Monkey IL-22 Specifications: (Molecular Weight: 16.6 kDa) (Amino Acid Sequence: APVSSHCRLD KSNFQQPYIT NRTFMLAKEA SSADNNTDVR LIGEKLFRGV SMSERCYLMK QVLNFTLEEV LLPQSDRFQP YMQEVVPFLA RLSNSLSTCH IEGDDLHIQK NVQKLKDTVK KLGESGEIKA IGELDLLFMS LRNACI (146)) (Gene ID: 102129310). For research use only. Made in the USA
Catalog No. 50-253-3433