Learn More
Kingfisher Biotech Inc Cynomolgus Monkey IL-4 Recombinant Protein (Yeast, Endotoxin - Free) - 5 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1184Y005
Encompass
The Cynomolgus Monkey IL-4 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Cynomolgus Monkey IL-4 applications are for cell culture, ELISA standard, and Western Blot Control. Cynomolgus Monkey IL-4 yeast-derived recombinant protein can be purchased in multiple sizes. Cynomolgus Monkey IL-4 Specifications: (Molecular Weight: 14.9 kDa) (Amino Acid Sequence: HKCDITLQEI IKTLNSLTEQ KTLCTKLTIT DILAASKNTT EKETFCRAAT VLRQFYSHHE KDTRCLGATA QQFHRHKQLI RFLKRLDRNL WGLAGLNSCP VKEANQSTLE NFLERLKTIM REKYSKCSS (129)) (Gene ID: 107130068). For research use only. Made in the USA
Catalog No. 50-253-3572