Learn More
Kingfisher Biotech Inc Cynomolgus Monkey IL-8 Recombinant Protein (Yeast, Endotoxin - Free) - 100 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1321Y100
Encompass
The Cynomolgus Monkey IL-8 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Cynomolgus Monkey IL-8 applications are for cell culture, ELISA standard, and Western Blot Control. Cynomolgus Monkey IL-8 yeast-derived recombinant protein can be purchased in multiple sizes. Cynomolgus Monkey IL-8 Specifications: (Molecular Weight: 9.3 kDa) (Amino Acid Sequence: EGAVLPRSAK ELRCECIKTY SKPFHPKFIK ELRVIESGPH CANTEIIVKL SDGRELCLDP KEPWVQRVVE KFVKRAENQN P (81)) (Gene ID: 102127272). For research use only. Made in the USA
Catalog No. 50-253-3398