Learn More
Cayman Chemical Cynomolgus Monkey Pregnane X Receptor Reporter Assay Kit, 96 Well
SDP
Catalog No. 502994309
Encompass_Preferred
cPXR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields exceptional cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for cumbersome intermediate treatment steps such as spin-and-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup. [INDIGO Catalog Nos. C07001-32, C07001]
Catalog No. 50-299-4309 Supplier Cayman Chemical Supplier No. 2140396 WELL
cPXR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields exceptional cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for cumbersome intermediate treatment steps such as spin-and-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup. [INDIGO Catalog Nos. C07001-32, C07001]