Learn More
Cayman Chemical Cynomolgus Monkey Peroxisome Proliferator-Activated Receptor Delta Reporter Assay System, 96 Well
SDP
Catalog No. 502939340
Encompass_Preferred
cPPARδ Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog Nos. C00121-32, C00121]
Catalog No. 50-293-9340 Supplier Cayman Chemical Supplier No. 2831096 WELL
cPPARδ Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog Nos. C00121-32, C00121]