Learn More
Cayman Chemical Cynomolgus Monkey Farnesoid X Receptor Reporter Assay System, 3x32 Well
SDP
Catalog No. 502988332
Encompass_Preferred
Cyn Monkey FXR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. C00601-32, C00601]
Catalog No. 50-298-8332 Supplier Cayman Chemical Supplier No. 302573X32 WELL
Cyn Monkey FXR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. C00601-32, C00601]