Learn More
Cayman Chemical Cynomolgus Monkey Peroxisome Proliferator-Activated Receptor Alpha Reporter Assay System, 3x32 Well
SDP
Catalog No. 502941534
Encompass_Preferred
cPPARα Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. C00111-32, C00111]
Catalog No. 50-294-1534 Supplier Cayman Chemical Supplier No. 283093X32 WELL
cPPARα Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. C00111-32, C00111]