Learn More
Sino Biological Recombinant Cynomolgus Mesothelin Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 91009C08H1
Encompass_Preferred
A DNA sequence encoding the Cynomolgus MSLN (XM_005590816.3) (Asp296-Gly580) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-241-1135