Learn More
ACROBiosystems Cynomolgus Mucin-1 / MUC-1 Protein, Fc Tag
Catalog No. 502054942
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus Mucin-1, Fc Tag (MU1-C5252) is expressed from human 293 cells (HEK293). It contains AA Leu 254 - Gly 373 (Accession # XP_005541632.1).;This protein carries a human IgG1 Fc tag at the C-terminus.The mature Mucin-1 is a non-covalent heterodimeric complex with the proteolytically cleaved N-terminal fragment and C-terminal fragment. Each N-terminal fragment and C-terminal fragment has a calculated MW of 7.2 kDa and 32.8 kDa. The protein migrates as 48 kDa (precursor protein) and 38-43 kDa (C-terminus) under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-205-4942 Supplier ACROBiosystems Supplier No. MU1C52521MG
Recombinant Protein;1MG;Cynomolgus Mucin-1, Fc Tag (MU1-C5252) is expressed from human 293 cells (HEK293). It contains AA Leu 254 - Gly 373 (Accession # XP_005541632.1).;This protein carries a human IgG1 Fc tag at the C-terminus.The mature Mucin-1 is a non-covalent heterodimeric complex with the proteolytically cleaved N-terminal fragment and C-terminal fragment. Each N-terminal fragment and C-terminal fragment has a calculated MW of 7.2 kDa and 32.8 kDa. The protein migrates as 48 kDa (precursor protein) and 38-43 kDa (C-terminus) under reducing (R) condition (SDS-PAGE) due to glycosylation.