Learn More
ACROBiosystems Cynomolgus Nectin-4 25ug
Catalog No. 502534389
Encompass_Preferred
Biotinylated Cynomolgus Nectin-4 Protein HisAvitag (NE4-C82E5) is expressed from human 293 cells (HEK293). It contains AA Gly 32 - Ser 349 (Accession # XP_005541277.1).
Catalog No. 50-253-4389 Supplier ACROBiosystems Supplier No. NE4C82E525UG
Biotinylated Cynomolgus Nectin-4 Protein HisAvitag (NE4-C82E5) is expressed from human 293 cells (HEK293). It contains AA Gly 32 - Ser 349 (Accession # XP_005541277.1).