Learn More
Sino Biological Cynomolgus NECTIN2 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 90206C08H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus (NP_001037058.1) (Met1-Leu360) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-3422