Learn More
Sino Biological Recombinant Cynomolgus Nectin-4 Protein (ECD, hFc Tag)
Supplier:  Sino Biological 90957C02H1
Encompass_Preferred
A DNA sequence encoding the Cynomolgus NECTIN4 (XP_005541277.1) (Met1-Ser349) was expressed with the fused Fc region of human IgG1 at the C-terminus.
Catalog No. 50-224-6566