Learn More
ACROBiosystems ACROBiosystems Cynomolgus NKG2A / CD159a Protein, His Tag
Catalog No. 502106429
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus NKG2A, His Tag (NKA-C5245) is expressed from human 293 cells (HEK293). It contains AA Pro 94 - Leu 233 (Accession # Q68VD2-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 17.9 kDa. The protein migrates as 27-37 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6429 Supplier ACROBiosystems Supplier No. NKAC5245100UG
Recombinant Protein;100UG;Cynomolgus NKG2A, His Tag (NKA-C5245) is expressed from human 293 cells (HEK293). It contains AA Pro 94 - Leu 233 (Accession # Q68VD2-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 17.9 kDa. The protein migrates as 27-37 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.