Learn More
ACROBiosystems Cynomolgus NKG2A&CD94, His Tag (NC4-C52H6) is expressed from human 293 cells (HEK293). It contains AA Ala 113 - Leu 233 (NKG2A) & Asp 57 - Ile 179 (CD94) (Accession # Q68VD2-1 (NKG2A) & Q68VD4-1 (CD94)).
Catalog No. 502231491
Encompass_Preferred
CD94 plays a role as a receptor for the recognition of MHC class I HLA-E molecules by NK cells and some cytotoxic T-cells. KLRD1 (CD94) is an antigen preferentially expressed on NK cells and is classified as a type II membrane protein because it has an external C terminus. NKG2A/CD159a is a transmembrane protein belonging to the CD94/NKG2 family of C-type lectin-like receptors that inhibits innate immune system activation. CD94 pairs with the NKG2 molecule as a heterodimer. The CD94/NKG2 complex, on the surface of natural killer cells interacts with Human Leukocyte Antigen (HLA)-E on target cells.
This item is not returnable. View return policy
Catalog No. 50-223-1491 Supplier ACROBiosystems Supplier No. NC4C52H6100UG
CD94 plays a role as a receptor for the recognition of MHC class I HLA-E molecules by NK cells and some cytotoxic T-cells. KLRD1 (CD94) is an antigen preferentially expressed on NK cells and is classified as a type II membrane protein because it has an external C terminus. NKG2A/CD159a is a transmembrane protein belonging to the CD94/NKG2 family of C-type lectin-like receptors that inhibits innate immune system activation. CD94 pairs with the NKG2 molecule as a heterodimer. The CD94/NKG2 complex, on the surface of natural killer cells interacts with Human Leukocyte Antigen (HLA)-E on target cells.