Learn More
ACROBiosystems Cynomolgus NKG2C / CD159c Protein, His Tag
Catalog No. 502055237
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus NKG2C, His Tag (NKC-C5247) is expressed from human 293 cells (HEK293). It contains AA Ile 94 - Leu 231 (Accession # Q68VD0-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 17.8 kDa. The protein migrates as 25 kDa, 27-30 kDa and 35-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-205-5237 Supplier ACROBiosystems Supplier No. NKCC52471MG
Recombinant Protein;1MG;Cynomolgus NKG2C, His Tag (NKC-C5247) is expressed from human 293 cells (HEK293). It contains AA Ile 94 - Leu 231 (Accession # Q68VD0-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 17.8 kDa. The protein migrates as 25 kDa, 27-30 kDa and 35-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.