Learn More
ACROBiosystems ACROBiosystems Cynomolgus NKp46 / NCR1 / CD335 Protein, His Tag (MALS verified)
Catalog No. 502106530
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus NKp46, His Tag (NC1-C52H4) is expressed from human 293 cells (HEK293). It contains AA Pro 22 - Arg 257 (Accession # Q95JB9-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.6 kDa. The protein migrates as 34-40 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6530 Supplier ACROBiosystems Supplier No. NC1C52H41MG
Recombinant Protein;1MG;Cynomolgus NKp46, His Tag (NC1-C52H4) is expressed from human 293 cells (HEK293). It contains AA Pro 22 - Arg 257 (Accession # Q95JB9-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 28.6 kDa. The protein migrates as 34-40 KDa under reducing (R) condition (SDS-PAGE) due to glycosylation.