Learn More
ACROBiosystems ACROBiosystems Cynomolgus NKp46 / NCR1 / CD335 Protein, Llama IgG2b Fc Tag, low endotoxin
Supplier:  ACROBiosystems NC1C52551MG
Encompass_Preferred
Recombinant Protein;1MG;Cynomolgus NKp46, Llama IgG2b Fc Tag (NC1-C5255) is expressed from human 293 cells (HEK293). It contains AA Pro 22 - Arg 257 (Accession # Q95JB9-1).;This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 54.7 kDa. The protein migrates as 60-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus NKp46, Llama IgG2b Fc Tag (Cat. No. NC1-C5255) at 1 μg/mL (100 μL/well) can bind Anti-NKp46, hIgG4 with a linear range of 0.4-6 ng/mL (QC tested).
Catalog No. 50-210-6563